Learn More
1-Lauroyl-2-myristoyl-3-chloropropanediol, TRC
Product Code. 30137279
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137279 100 mg 100mg
30007097 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137279 Supplier Toronto Research Chemicals Supplier No. TRCL178700100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2828775-74-0
Molecular Formula C29H55ClO4
Molecular Weight (g/mol) 503.2
IUPAC Name (S)-1-chloro-3-(dodecanoyloxy)propan-2-yl tetradecanoate
SMILES O=C(CCCCCCCCCCCCC)O[C@H](CCl)COC(CCCCCCCCCCC)=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C29H55ClO4/c1-3-5-7-9-11-13-14-16-18-20-22-24-29(32)34-27(25-30)26-33-28(31)23-21-19-17-15-12-10-8-6-4-2/h27H,3-26H2,1-2H3/t27-/m1/s1
Formula Weight 502.38
Chemical Name or Material 1-Lauroyl-2-myristoyl-3-chloropropanediol