Learn More
1-Isopropyl-3-(4-fluorophenyl)indole-2-carboxaldehyde, TRC
Product Code. 30403111
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403111 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30403111 Supplier Toronto Research Chemicals Supplier No. TRCI8245301G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Isopropyl-3-(4-fluorophenyl)indole-2-carboxaldehyde
CAS 101125-34-2
Molecular Formula C18 H16 F N O
Quantity 1 g
Synonym 1H-Indole-2-carboxaldehyde, 3-(4-fluorophenyl)-1-(1-methylethyl)-, 3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indole-2-carboxaldehyde, 3-(4-Fluorophenyl)-1-isopropyl-1H-indole-2-carbaldehyde, 3-(4-Fluorophenyl)-1-isopropyl-1H-indole-2-carboxaldehyde
InChI Formula InChI=1S/C18H16FNO/c1-12(2)20-16-6-4-3-5-15(16)18(17(20)11-21)13-7-9-14(19)10-8-13/h3-12H,1-2H3
SMILES CC(C)n1c(C=O)c(c2ccc(F)cc2)c3ccccc13
IUPAC Name 3-(4-fluorophenyl)-1-propan-2-ylindole-2-carbaldehyde
Molecular Weight (g/mol) 281.32
Formula Weight 281.1216