Learn More
1-Isopropyl-3-(4-fluorophenyl)indole-2-carboxaldehyde, TRC
Product Code. 30389092
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389092 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30389092 Supplier Toronto Research Chemicals Supplier No. TRCI824530250MG
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material 1-Isopropyl-3-(4-fluorophenyl)indole-2-carboxaldehyde
CAS 101125-34-2
Molecular Formula C18 H16 F N O
Quantity 250 mg
Synonym 1H-Indole-2-carboxaldehyde, 3-(4-fluorophenyl)-1-(1-methylethyl)-, 3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indole-2-carboxaldehyde, 3-(4-Fluorophenyl)-1-isopropyl-1H-indole-2-carbaldehyde, 3-(4-Fluorophenyl)-1-isopropyl-1H-indole-2-carboxaldehyde
InChI Formula InChI=1S/C18H16FNO/c1-12(2)20-16-6-4-3-5-15(16)18(17(20)11-21)13-7-9-14(19)10-8-13/h3-12H,1-2H3
SMILES CC(C)n1c(C=O)c(c2ccc(F)cc2)c3ccccc13
IUPAC Name 3-(4-fluorophenyl)-1-propan-2-ylindole-2-carbaldehyde
Molecular Weight (g/mol) 281.32
Formula Weight 281.1216