Learn More
1-Iodopropane-1,1,3,3,3-d5, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Iodopropane-1,1,3,3,3-D5 |
| CAS | 1219794-94-1 |
| Molecular Formula | C3 D5 H2 I |
| Quantity | 0.25 g |
| Synonym | 3-Iodopropane-1,1,1,3,3-d5, 1-Iodopropane-1,1,3,3,3-d5, 1-Iodopropane-d5, 1-Iodopropane-1,1,3,3,3-D5, [1,1,3,3,3-2H5]Propyl iodide |
| InChI Formula | InChI=1 S/C3H7I/c1-2-3-4/h2-3H2,1H3/i1D3,3D2 |
| SMILES | [2 H]C([2 H])([2 H])CC([2 H])([2 H])I |
| IUPAC Name | 1,1,1,3,3-pentadeuterio-3-iodopropane |
| Molecular Weight (g/mol) | 175.023 |
| Formula Weight | 174.9906 g/mol |
| Show More |