missing translation for 'onlineSavingsMsg'
Learn More

1-Iodododecane, 98%, stabilized

Product Code. 10075663 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 10075663

Brand: Thermo Scientific Acros 250091000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4292-19-7 | C12H25I | 296.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4292-19-7
Molecular Formula C12H25I
Molecular Weight (g/mol) 296.24
MDL Number MFCD00001088
InChI Key GCDPERPXPREHJF-UHFFFAOYSA-N
Synonym n-dodecyl iodide, dodecyl iodide, lauryl iodide, dodecane, 1-iodo, dodecane,1-iodo, iodododecane, iodo-dodecane, 1-iodanyldodecane, pubchem3943, n-iodododecane
PubChem CID 20282
IUPAC Name 1-iodododecane
SMILES CCCCCCCCCCCCI

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -3.0°C
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 159.0°C to 160.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)11I
Refractive Index 1.4830 to 1.4850
Quantity 100 mL
Beilstein 01,67
Specific Gravity 1.2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 296.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iodododecane, Stabilized with 500ppm copper
Show More Show Less
compliance-icons
Product Identifier
  • 1-Iodododecane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.