Learn More
1-(hydroxymethyl)cyclopentan-1-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(hydroxymethyl)cyclopentan-1-ol |
| CAS | 74397-18-5 |
| Molecular Formula | C6H12O2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C6H12O2/c7-5-6(8)3-1-2-4-6/h7-8H,1-5H2 |
| SMILES | OCC1(O)CCCC1 |
| IUPAC Name | 1-(hydroxymethyl)cyclopentan-1-ol |
| Molecular Weight (g/mol) | 116.15 |
| Formula Weight | 116.0837 |