Learn More
1-Hydroxy-3,3-dimethylbutan-2-one, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Hydroxy-3,3-dimethylbutan-2-one |
| CAS | 38895-88-4 |
| Molecular Formula | C6H12O2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C6H12O2/c1-6(2,3)5(8)4-7/h7H,4H2,1-3H3 |
| SMILES | CC(C)(C)C(=O)CO |
| IUPAC Name | 1-hydroxy-3,3-dimethylbutan-2-one |
| Molecular Weight (g/mol) | 116.15 |
| Formula Weight | 116.0837 |