Learn More
(1-Hexadecyl)trimethylammonium chloride, 96%
Description
(1-Hexadecyl)trimethylammonium chloride is used as a topical antiseptic and surfactant. It is used in hair care formulations, cream rinse ingredient, hair conditioners and shampoos as a conditioning agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 234°C |
| Color | White |
| Density | 0.968 g/mL |
| Odor | Odorless |
| Linear Formula | CH3(CH2)15N(CH3)3Cl |
| Quantity | 25 g |
| UN Number | UN1759 |
| Beilstein | 3657974 |
| Solubility Information | Soluble in water. |
| Formula Weight | 320 |
| Show More |
Safety and Handling
- (1-Hexadecyl)trimethylammonium chloride
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : (1-Hexadecyl)trimethylammonium chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.