missing translation for 'onlineSavingsMsg'
Learn More

1-Heptadecyl Bromide, TRC

Product Code. 30400296
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400296 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400296 Supplier Toronto Research Chemicals Supplier No. TRCH2790001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Heptadecyl Bromide
CAS 3508-00-7
Molecular Formula C17 H35 Br
Quantity 1 g
InChI Formula InChI=1S/C17H35Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18/h2-17H2,1H3
SMILES CCCCCCCCCCCCCCCCCBr
IUPAC Name 1-bromoheptadecane
Molecular Weight (g/mol) 319.36
Formula Weight 318.1922
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.