Learn More
1-Eicosanethiol, 98%
Product Code. 15487447
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487447 1 g 1g
15497447 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15487447 Supplier Thermo Scientific Alfa Aesar Supplier No. H34498.03
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
CAS: 13373-97-2 | C20H42S | 314.616 g/mol
1-Eicosanethiol is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Eicosanethiol is used as an organic chemical synthesis intermediate.
Notes
Air and moisture sensitive. Incompatible with heat.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13373-97-2
Molecular Formula C20H42S
Molecular Weight (g/mol) 314.616
MDL Number MFCD07779406
InChI Key YYHYWOPDNMFEAV-UHFFFAOYSA-N
Synonym 1-eicosanethiol, eicosanethiol, 1-icosanethiol, icosyl mercaptan, eicosyl mercaptan, acmc-209btf
PubChem CID 139443
IUPAC Name icosane-1-thiol
SMILES CCCCCCCCCCCCCCCCCCCCS
Specifications
Melting Point 38°C to 41°C
Odor Stench
Linear Formula CH3(CH2)19SH
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 314.61
Percent Purity 98%
Chemical Name or Material 1-Eicosanethiol