Learn More
1-Cyclopropyl-8-ethoxy-6,7-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Cyclopropyl-8-ethoxy-6,7-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid |
| CAS | 143158-55-8 |
| Molecular Formula | C15H13F2NO4 |
| Quantity | 50 mg |
| Synonym | 3-Quinolinecarboxylic acid, 1-cyclopropyl-8-ethoxy-6,7-difluoro-1,4-dihydro-4-oxo-, 1-cyclopropyl-8-ethoxy-6,7-difluoro-4-oxo-quinoline-3-carboxylic acid |
| InChI Formula | InChI=1S/C15H13F2NO4/c1-2-22-14-11(17)10(16)5-8-12(14)18(7-3-4-7)6-9(13(8)19)15(20)21/h5-7H,2-4H2,1H3,(H,20,21) |
| SMILES | CCOc1c(F)c(F)cc2C(=O)C(=CN(C3CC3)c12)C(=O)O |
| IUPAC Name | 1-cyclopropyl-8-ethoxy-6,7-difluoro-4-oxoquinoline-3-carboxylic acid |
| Molecular Weight (g/mol) | 309.26 |
| Formula Weight | 309.0813 |