Learn More
1-Cyclopentenecarboxylic Acid, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 1-Cyclopentenecarboxylic Acid |
| CAS | 1560-11-8 |
| Molecular Formula | C6H8O2 |
| Quantity | 100 mg |
| Synonym | Isoaleprolic Acid, NSC 44968 |
| InChI Formula | InChI=1S/C6H8O2/c7-6(8)5-3-1-2-4-5/h3H,1-2,4H2,(H,7,8) |
| SMILES | OC(=O)C1=CCCC1 |
| IUPAC Name | cyclopentene-1-carboxylic acid |
| Molecular Weight (g/mol) | 112.13 |
| Formula Weight | 112.0524 |