Learn More
1-Cyclohexylnaphthalene, TRC
Product Code. 30366897
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366897 1 g -
30349756 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30366897 Supplier Toronto Research Chemicals Supplier No. TRCC9880551G
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3042-69-1
Molecular Formula C16H18
Molecular Weight (g/mol) 210.31
Synonym 1-Cyclohexyl-naphthalene, NSC 131573
IUPAC Name 1-cyclohexylnaphthalene
SMILES C1CCC(CC1)C2=CC=CC3=CC=CC=C32
Specifications
Quantity 1 g
InChI Formula InChI=1S/C16H18/c1-2-7-13(8-3-1)16-12-6-10-14-9-4-5-11-15(14)16/h4-6,9-13H,1-3,7-8H2
Formula Weight 210.1409
Chemical Name or Material 1-Cyclohexylnaphthalene