missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-1-tosylmethyl Isocyanide, TRC

Product Code. 30364404
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364404 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30364404 Supplier Toronto Research Chemicals Supplier No. TRCC986260500MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Cyclohexyl-1-tosylmethyl Isocyanide
CAS 1048971-67-0
Molecular Formula C15 H19 N O2 S
Quantity 500 mg
Synonym 1-[(Cyclohexylisocyanomethyl)sulfonyl]-4-methylbenzene
InChI Formula InChI=1S/C15H19NO2S/c1-12-8-10-14(11-9-12)19(17,18)15(16-2)13-6-4-3-5-7-13/h8-11,13,15H,3-7H2,1H3
SMILES Cc1ccc(cc1)S(=O)(=O)C([N+]#[C-])C2CCCCC2
IUPAC Name 1-[cyclohexyl(isocyano)methyl]sulfonyl-4-methylbenzene
Molecular Weight (g/mol) 277.382
Formula Weight 277.114
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.