missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloropentane, 98%, ACROS Organics™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | -60°C |
| Color | Colorless |
| Density | 0.8800g/mL |
| Boiling Point | 107°C to 108°C |
| Flash Point | 11°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
Product Identifier
- 1-Chloropentane
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 2
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction