missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorobutane-d9, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Chlorobutane-D9 |
| CAS | 175540-76-8 |
| Molecular Formula | C4 D9 Cl |
| Quantity | 0.25 g |
| Synonym | Butane-1,1,1,2,2,3,3,4,4-d9, 4-chloro- (9 CI), 4-Chlorobutane-1,1,1,2,2,3,3,4,4-d9, 1-Chlorobutane-d9, 1-Chlorobutane-D9 |
| InChI Formula | InChI=1 S/C4H9Cl/c1-2-3-4-5/h2-4H2,1H3/i1D3,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Cl |
| IUPAC Name | 1-chloro-1,1,2,2,3,3,4,4,4-nonadeuteriobutane |
| Molecular Weight (g/mol) | 101.6227 |
| Formula Weight | 101.0958 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction