Learn More
1-Chloro-3-[(1Z)-1-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-1-propen-1-yl]-benzene, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Chloro-3-((1Z)-1-(((1,1-dimethylethyl)dimethylsilyl)oxy)-1-propen-1-yl)-benzene |
| CAS | 291275-45-1 |
| Molecular Formula | C15H23ClOSi |
| Quantity | 1 g |
| Synonym | [[(1Z)-1-(3-Chlorophenyl)-1-propenyl]oxy](1,1-dimethylethyl)dimethyl-silane, 1-Chloro-3-[(1Z)-1-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-1-propen-1-yl]benzene |
| InChI Formula | InChI=1S/C15H23ClOSi/c1-7-14(12-9-8-10-13(16)11-12)17-18(5,6)15(2,3)4/h7-11H,1-6H3/b14-7- |
| SMILES | C\C=C(/O[Si](C)(C)C(C)(C)C)\c1cccc(Cl)c1 |
| IUPAC Name | tert-butyl-[(Z)-1-(3-chlorophenyl)prop-1-enoxy]-dimethylsilane |
| Molecular Weight (g/mol) | 282.88 |
| Formula Weight | 282.1207 |