Learn More
1-Chloro-2,4-dinitrobenzene-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Chloro-2,4-dinitrobenzene-D3 |
| CAS | 347840-12-4 |
| Molecular Formula | C6 D3 Cl N2 O4 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2,4-d3, 6-chloro-3,5-dinitro- (9 CI), 1-Chloro-2,4-dinitrobenzene-d3, 1-Chloro-2,4-dinitrobenzene-3,5,6-d3, 6-Chloro-3,5-dinitrobenzene-1,2,4-d3, 1-Chloro-2,4-dinitrobenzene-D3 |
| InChI Formula | InChI=1 S/C6H3ClN2O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3 H/i1D,2 D,3 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c(c1Cl)[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | 1-chloro-2,3,5-trideuterio-4,6-dinitrobenzene |
| Molecular Weight (g/mol) | 205.5705 |
| Formula Weight | 204.997 g/mol |
| Show More |