Learn More
1-Chloro-1,2-benziodoxol-3(1H)-one, TRC
Product Code. 30367400
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367400 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30367400 Supplier Toronto Research Chemicals Supplier No. TRCC363558250MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Chloro-1,2-benziodoxol-3(1H)-one
CAS 59457-26-0
Molecular Formula C7H4ClIO2
Quantity 250 mg
Synonym 1-Chloro-1,2-benziodoxol-3-one, 1-Chlorobenziodoxol-3-one, 3-Oxo-3H-2,1-benzoxiodolium Chloride
InChI Formula InChI=1S/C7H4ClIO2/c8-9-6-4-2-1-3-5(6)7(10)11-9/h1-4H
SMILES ClI1OC(=O)c2ccccc12
IUPAC Name 1-chloro-1λ3,2-benziodoxol-3-one
Molecular Weight (g/mol) 282.46