Learn More
1-Butyl-3-(1,1-dioxido-2H-1,2,4-benzothiadiazin-3-yl)-4-hydroxy-2(1H)-quinolinone, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Butyl-3-(1,1-dioxido-2H-1,2,4-benzothiadiazin-3-yl)-4-hydroxy-2(1H)-quinolinone |
| CAS | 303776-83-2 |
| Molecular Formula | C20H19N3O4S |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C20H19N3O4S/c1-2-3-12-23-15-10-6-4-8-13(15)18(24)17(20(23)25)19-21-14-9-5-7-11-16(14)28(26,27)22-19/h4-11,24H,2-3,12H2,1H3,(H,21,22) |
| SMILES | CCCCN1C(=O)\C(=C\2/Nc3ccccc3S(=O)(=O)N2)\C(=O)c4ccccc14 |
| IUPAC Name | 1-butyl-3-(1,1-dioxo-4H-1λ6,2,4-benzothiadiazin-3-yl)-4-hydroxyquinolin-2-one |
| Molecular Weight (g/mol) | 397.45 |
| Formula Weight | 397.1096 |