Learn More
1-Bromotridecane-1,1,2,2-d4, CDN
Product Code. 30318814
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318814 0.1 g 0.1g
30312132 0.25 g 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318814 Supplier CDN Supplier No. CDND64600.1G
 Restricted Item
Estimated Shipment: 10-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromotridecane-1,1,2,2-D4
CAS 284474-45-9
Molecular Formula C13 D4 H23 Br
Quantity 0.1 g
Synonym Tridecane-1,1,2,2-d4, 1-bromo- (9 CI), 1-Bromotridecane-1,1,2,2-d4, 1-Bromotridecane-1,1,2,2-D4, Tridecyl-1,1,2,2-d4 bromide
InChI Formula InChI=1 S/C13H27Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14/h2-13H2,1H3/i12D2,13D2
SMILES [2 H]C([2 H])(Br)C([2 H])([2 H])CCCCCCCCCCC
IUPAC Name 1-bromo-1,1,2,2-tetradeuteriotridecane
Molecular Weight (g/mol) 267.2821
Formula Weight 266.1547 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity