Learn More
1-Bromohexane-1,1-d2, CDN
Product Code. 30327384
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30327384
Brand: CDN CDND71640.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromohexane-1,1-D2
CAS 78904-38-8
Molecular Formula C6 D2 H11 Br
Quantity 0.5 g
Synonym Hexane-1,1-d2, 1-bromo- (9 CI, ACI), 1-Bromohexane-1,1-d2 (ACI), 1-Bromohexane-1,1-D2, 1-Bromohexane-d2
InChI Formula InChI=1 S/C6H13Br/c1-2-3-4-5-6-7/h2-6H2,1H3/i6D2
SMILES [2 H]C([2 H])(Br)CCCCC
IUPAC Name 1-bromo-1,1-dideuteriohexane
Molecular Weight (g/mol) 167.0837
Formula Weight 166.0326 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity