Learn More
1-Bromo-6-chlorohexane, 97%
Product Code. 11417244
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11417244 5 g 5g
11427244 25 g 25g
2 options
Product Code. 11417244
Brand: Thermo Scientific Alfa Aesar B21619.06
Request Bulk or Custom Format
CAS: 6294-17-3 | C6H12BrCl | 199.52 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6294-17-3
Molecular Formula C6H12BrCl
Molecular Weight (g/mol) 199.52
MDL Number MFCD00001019
InChI Key JTYUIAOHIYZBPB-UHFFFAOYSA-N
Synonym 6-chlorohexyl bromide, hexane, 1-bromo-6-chloro, 1-chloro-6-bromohexane, hexyl bromide, 6-chloro, 6-bromohexyl chloride, 1-bromo-6-chloro hexane, 1-bromo-6-chloro-hexane, acmc-1b9m0, 1-bromo-6-chlorohexane, jtyuiaohiyzbpb-uhfffaoysa
PubChem CID 80516
IUPAC Name 1-bromo-6-chlorohexane
SMILES ClCCCCCCBr
Specifications
Density 1.337
Boiling Point 109°C to 110°C (2 mmHg)
Flash Point 101°C (214°F)
Refractive Index 1.4806
Quantity 5 g
Formula Weight 199.53
Percent Purity 97%
Chemical Name or Material 1-Bromo-6-chlorohexane