missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(phenoxymethyl)benzene, 97%, Thermo Scientific™

Product Code. 10091974
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091974 1 g Amber Glass Bottle 1g
10235904 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10091974 Supplier Thermo Scientific Maybridge Supplier No. CC63710DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 20600-22-0
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
MDL Number MFCD00017847
InChI Key KZVLWVAXLYUCLW-UHFFFAOYSA-N
Synonym 1-bromo-4-phenoxymethyl benzene, 4-bromophenyl methoxy benzene, 4-bromobenzylphenyl ether, 4-bromobenzyl phenyl ether, 1-bromo4-phenoxymethyl-benzene, 1-bromo-4-phenoxymethyl-benzene, benzyl phenyl ether analogue, 6c
PubChem CID 726149
IUPAC Name 1-bromo-4-(phenoxymethyl)benzene
SMILES C1=CC=C(C=C1)OCC2=CC=C(C=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 263.13
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-(phenoxymethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.