Learn More
1-Bromo-4-chlorobenzene, 98+%
Product Code. 11430143
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
Product Code. 11430143
Brand: Thermo Scientific Alfa Aesar A12160.0B
Request Bulk or Custom Format
CAS: 106-39-8 | C6H4BrCl | 191.45 g/mol
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.
Solubility
Soluble in water 0.045g/L.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106-39-8
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.45
MDL Number MFCD00000600
InChI Key NHDODQWIKUYWMW-UHFFFAOYSA-N
Synonym 4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
PubChem CID 7806
IUPAC Name 1-bromo-4-chlorobenzene
SMILES ClC1=CC=C(Br)C=C1
Specifications
Melting Point 63°C to 67°C
Density 1.651
Boiling Point 196°C
Quantity 1000 g
Odor Aromatic
UN Number UN2811
Beilstein 1099014
Solubility Information Soluble in water 0.045g/L.
Formula Weight 191.46
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-4-chlorobenzene