Learn More
1-Bromo-3,5-difluorobenzene-d3, CDN
Product Code. 30317784
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317784
Brand: CDN CDND64750.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromo-3,5-difluorobenzene-D3
CAS 1219798-73-8
Molecular Formula C6 D3 Br F2
Quantity 0.5 g
Synonym 1-Bromo-3,5-difluorobenzene-d3, 1-Bromo-3,5-difluorobenzene-2,4,6-d3
InChI Formula InChI=1 S/C6H3BrF2/c7-4-1-5(8)3-6(9)2-4/h1-3 H/i1D,2 D,3 D
SMILES [2 H]c1c(F)c([2 H])c(Br)c([2 H])c1F
IUPAC Name 1-bromo-2,4,6-trideuterio-3,5-difluorobenzene
Molecular Weight (g/mol) 196.0073
Formula Weight 194.9574 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity