missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-methylpropane-3,3,3-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-2-methylpropane-3,3,3-D3 |
| CAS | 344299-42-9 |
| Molecular Formula | C4 D3 H6 Br |
| Quantity | 0.25 g |
| Synonym | Propane-1,1,1-d3, 3-bromo-2-methyl- (9 CI), 2-(Bromomethyl)-1,1,1-trideuteriopropane, 1-Bromo-2-methylpropane-3,3,3-d3, Isobutyl-d3 bromide, 1-Bromo-2-methylpropane-3,3,3-D3, 1-Bromo-2-methylpropane-d3, 1-Bromo-2-(methyl-d3)propane |
| InChI Formula | InChI=1 S/C4H9Br/c1-4(2)3-5/h4H,3H2,1-2H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(C)CBr |
| IUPAC Name | 3-bromo-1,1,1-trideuterio-2-methylpropane |
| Molecular Weight (g/mol) | 140.0368 |
| Formula Weight | 139.0076 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction