Learn More
1-Bromo-2-methyl-d3-propane-2,3,3,3-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-2-methylpropane-d7 |
| CAS | 344299-41-8 |
| Molecular Formula | C4 D7 H2 Br |
| Quantity | 0.5 g |
| Synonym | 1-Methyl-2-propyl Bromide-d7, 2-Methylpropyl Bromide-d7, Bromoisobutane-d7, Isobutyl Bromide-d7, NSC 8416-d7, iso-Butyl Bromide-d7 |
| InChI Formula | InChI=1 S/C4H9Br/c1-4(2)3-5/h4H,3H2,1-2H3/i1D3,2D3,4 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(CBr)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-(bromomethyl)-1,1,1,2,3,3,3-heptadeuteriopropane |
| Molecular Weight (g/mol) | 144.06 |
| Formula Weight | 143.0327 g/mol |
| Show More |