missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2,3-difluorobenzene, 98%, ACROS Organics™
Shop All Thermo Scientific Chemicals Products
Click to view available options
:
1g; Glass bottle
Specifications
Specifications
| CAS | 38573-88-5 |
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Color | Colorless to Peach |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Molecular Formula | C6H3BrF2 |
| Linear Formula | BrC6H3F2 |
| Refractive Index | 1.5090 to 1.5100 (20°C, 589nm) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction