missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2,3,5-trichlorobenzene-d2, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-2,3,5-trichlorobenzene-D2 |
| CAS | 1219803-86-7 |
| Molecular Formula | C6 D2 Br Cl3 |
| Quantity | 0.5 g |
| Synonym | 1-Bromo-2,3,5-trichlorobenzene-d2, 1-Bromo-3,5,6-trichlorobenzene-2,4-d2 |
| InChI Formula | InChI=1 S/C6H2BrCl3/c7-4-1-3(8)2-5(9)6(4)10/h1-2 H/i1D,2 D |
| SMILES | [2 H]c1c(Cl)c([2 H])c(Br)c(Cl)c1Cl |
| IUPAC Name | 1-bromo-2,3,5-trichloro-4,6-dideuteriobenzene |
| Molecular Weight (g/mol) | 262.3554 |
| Formula Weight | 259.8531 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction