missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-6-methoxyindole-2-boronic acid, 98%, Thermo Scientific™

Product Code. 15469227
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469227 1 g 1g
15479227 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469227 Supplier Thermo Scientific Alfa Aesar Supplier No. H52482.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-65-9
Molecular Formula C14H18BNO5
Molecular Weight (g/mol) 291.11
MDL Number MFCD06659830
InChI Key ATIJYIUOXDCYTN-UHFFFAOYSA-N
Synonym 1-boc-6-methoxyindole-2-boronic acid, 1-tert-butoxycarbonyl-6-methoxy-1h-indol-2-yl boronic acid, 6-methoxy-1h-indole-2-boronic acid, n-boc protected, 1-boc-6-methoxy-2-indoleboronic acid, 6-methoxyindole-2-boronic acid, boc protected, 1-tert-butoxycarbonyl-6-methoxyindol-2-ylboronic acid, 1-t-butoxycarbonyl-6-methoxyindole-2-boronic acid, 1h-indole-1-carboxylic acid, 2-borono-6-methoxy-, 1-1,1-dimethylethyl ester, 1h-indole-1-carboxylicacid, 2-borono-6-methoxy-, 1-1,1-dimethylethyl ester
PubChem CID 22309457
IUPAC Name [6-methoxy-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
SMILES B(C1=CC2=C(N1C(=O)OC(C)(C)C)C=C(C=C2)OC)(O)O

Specifications

Melting Point 100°C to 103°C
Quantity 1 g
Formula Weight 291.11
Percent Purity 98%
Chemical Name or Material 1-Boc-6-methoxyindole-2-boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.