Learn More
(+/-)-1-Boc-3-bromopyrrolidine, 95%
Product Code. 15463618
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463618 1 g 1g
15483618 250 mg 250mg
15473618 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15463618 Supplier Thermo Scientific Alfa Aesar Supplier No. H57071.03
Request Bulk or Custom Format
CAS: 939793-16-5 | C9H16BrNO2 | 250.14 g/mol
(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 939793-16-5
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.14
MDL Number MFCD08752478
InChI Key QJTKPXFJOXKUEY-UHFFFAOYNA-N
PubChem CID 42614217
IUPAC Name tert-butyl 3-bromopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(Br)C1
Specifications
Melting Point 45°C to 53°C
Quantity 1 g
UN Number UN3077
Synonym n-boc-3-bromopyrrolidine, 1-boc-3-bromopyrrolidine, 3-bromo-pyrrolidine-1-carboxylic acid tert-butyl ester, +/--1-boc-3-bromopyrrolidine, acmc-209zg9, ^+-1-boc-3-bromopyrrolidine, rs 1-tertbutoxycarbonyl-3-bromopyrrolidine, rs 1-tert-butoxycarbonyl-3-bromopyrrolidine, 2-methyl-2-propanyl 3-bromo-1-pyrrolidinecarboxylate
Solubility Information Slightly soluble in water.
Formula Weight 250.14
Percent Purity 95%
Chemical Name or Material (+/-)-1-Boc-3-bromopyrrolidine