Learn More
1-Benzylbenzimidazole, 97%, Thermo Scientific™
Product Code. 15466558
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15466558
Brand: Thermo Scientific Alfa Aesar H60088.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4981-92-4
Molecular Formula C14H21NO2S
Molecular Weight (g/mol) 267.387
MDL Number MFCD00458798
InChI Key PYBYEBRVJRDWPB-LBPRGKRZSA-N
Synonym s-2-benzyl-2-n-bocamino-ethyl thiol, s-tert-butyl 1-mercapto-3-phenylpropan-2-yl carbamate, tert-butyl n-2s-1-phenyl-3-sulfanylpropan-2-yl carbamate, s-2-benzyl-2-n-bocaminoethyl thiol, s-2-benzyl-2-n-boc-aminoethyl thiol, s-2-n-boc-2-amino-3-phenylpropane-1-thiol, s-1-mercaptomethyl-2-phenylethyl carbamic acid tert-butyl ester
PubChem CID 15003862
IUPAC Name tert-butyl N-[(2S)-1-phenyl-3-sulfanylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CS
Specifications
Melting Point 108°C to 110°C
Quantity 5 g
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material 1-Benzylbenzimidazole