missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-cyanoacetyl Urea, TRC

Product Code. 30344131
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344131 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344131 Supplier Toronto Research Chemicals Supplier No. TRCB2337501G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Benzyl-3-cyanoacetylurea
CAS 126245-46-3
Molecular Formula C11 H11 N3 O2
Quantity 1 g
InChI Formula InChI=1S/C11H11N3O2/c12-7-6-10(15)14-11(16)13-8-9-4-2-1-3-5-9/h1-5H,6,8H2,(H2,13,14,15,16)
SMILES O=C(CC#N)NC(=O)NCc1ccccc1
IUPAC Name N-(benzylcarbamoyl)-2-cyanoacetamide
Molecular Weight (g/mol) 217.22
Formula Weight 217.0851
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.