Learn More
1-Benzyl 2-Methyl (2R)-5-Oxopyrrolidine-1,2-dicarboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Benzyl 2-Methyl (2R)-5-Oxopyrrolidine-1,2-dicarboxylate |
| CAS | 690211-34-8 |
| Molecular Formula | C14H15NO5 |
| Quantity | 250 mg |
| Synonym | 1-Benzyl 2-Methyl (R)-5-Oxopyrrolidine-1,2-dicarboxylate |
| InChI Formula | InChI=1S/C14H15NO5/c1-19-13(17)11-7-8-12(16)15(11)14(18)20-9-10-5-3-2-4-6-10/h2-6,11H,7-9H2,1H3/t11-/m1/s1 |
| SMILES | COC(=O)[C@H]1CCC(=O)N1C(=O)OCc2ccccc2 |
| IUPAC Name | 1-O-benzyl 2-O-methyl (2R)-5-oxopyrrolidine-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 277.27 |
| Formula Weight | 277.095 |