Learn More
1'-Benzyl-1,4'-bipiperidine-4'-carbonitrile-d10, TRC
Product Code. 30344947
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344947 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30344947 Supplier Toronto Research Chemicals Supplier No. TRCB232982100MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1'-Benzyl-1,4'-bipiperidine-4'-carbonitrile-d10
CAS 1329624-69-2
Molecular Formula C18H15D10N3
Quantity 100 mg
Synonym 1'-(Phenylmethyl)-[1,4'-bipiperidine]-4'-carbonitrile-d10
InChI Formula InChI=1S/C18H25N3/c19-16-18(21-11-5-2-6-12-21)9-13-20(14-10-18)15-17-7-3-1-4-8-17/h1,3-4,7-8H,2,5-6,9-15H2/i2D2,5D2,6D2,11D2,12D2
SMILES [2H]C1([2H])N(C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H])C2(CCN(Cc3ccccc3)CC2)C#N
IUPAC Name 1-benzyl-4-(2,2,3,3,4,4,5,5,6,6-decadeuteriopiperidin-1-yl)piperidine-4-carbonitrile
Molecular Weight (g/mol) 293.47
Formula Weight 293.2676