Learn More
1'-Benzyl-1,4'-bipiperidine-4'-carbonitrile-d10, TRC
Specifications
Specifications
| Chemical Name or Material | 1'-Benzyl-1,4'-bipiperidine-4'-carbonitrile-d10 |
| CAS | 1329624-69-2 |
| Molecular Formula | C18H15D10N3 |
| Quantity | 10 mg |
| Synonym | 1'-(Phenylmethyl)-[1,4'-bipiperidine]-4'-carbonitrile-d10 |
| InChI Formula | InChI=1S/C18H25N3/c19-16-18(21-11-5-2-6-12-21)9-13-20(14-10-18)15-17-7-3-1-4-8-17/h1,3-4,7-8H,2,5-6,9-15H2/i2D2,5D2,6D2,11D2,12D2 |
| SMILES | [2H]C1([2H])N(C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H])C2(CCN(Cc3ccccc3)CC2)C#N |
| IUPAC Name | 1-benzyl-4-(2,2,3,3,4,4,5,5,6,6-decadeuteriopiperidin-1-yl)piperidine-4-carbonitrile |
| Molecular Weight (g/mol) | 293.47 |
| Formula Weight | 293.2676 |