Learn More
1-Benzyl-1,2,3-triazole-4,5-dicarboxylic acid, 98%, Thermo Scientific™
Product Code. 11348165
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348165 1 g 1g
11358165 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11348165 Supplier Thermo Scientific Alfa Aesar Supplier No. L08434.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 73953-89-6
Molecular Formula C11H9N3O4
Molecular Weight (g/mol) 247.21
MDL Number MFCD00043512
InChI Key IYYBUXUZYWGZGB-UHFFFAOYSA-N
Synonym 1-benzyl-1,2,3-triazole-4,5-dicarboxylic acid, 1-benzyl-1h-1,2,3-triazole-4,5-dicarboxylic acid, v-triazole-4,5-dicarboxylic acid, 1-benzyl, 1h-1,2,3-triazole-4,5-dicarboxylic acid, 1-phenylmethyl, 4-26-00-00965 beilstein handbook reference, 1-benzyl1,2,3-triazole-4,5-dicarboxylic acid, 1-benzyl-1,2,3-triazol-4,5-dicarboxylic acid, 1-benzyl-1h-1,2,3 triazole-4,5-dicarboxylic acid, 1-benzyl-1h-1,2,3-triazole-4,5-dicarboxylic acid #
PubChem CID 427031
IUPAC Name 1-benzyltriazole-4,5-dicarboxylic acid
SMILES C1=CC=C(C=C1)CN2C(=C(N=N2)C(=O)O)C(=O)O
Specifications
Melting Point 178°C to 182°C
Quantity 1 g
Beilstein 29417
Formula Weight 247.21
Percent Purity 98%
Chemical Name or Material 1-Benzyl-1,2,3-triazole-4,5-dicarboxylic acid