missing translation for 'onlineSavingsMsg'
Learn More

1-Allyl-3,7-dimethyl-8-p-sulfophenylxanthine Sodium Salt, TRC

Product Code. 30322870
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322870 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322870 Supplier Toronto Research Chemicals Supplier No. TRCA5526502.5MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Allyl-3,7-dimethyl-8-p-sulfophenylxanthine, Sodium Salt
CAS 149981-25-9
Molecular Formula C16 H15 N4 O5 S . Na
Quantity 2.5 mg
Synonym 1-Allyl-3,7-Dimethyl-8-P-Sulfophenylxanthine
InChI Formula InChI=1S/C16H16N4O5S.Na/c1-4-9-20-15(21)12-14(19(3)16(20)22)17-13(18(12)2)10-5-7-11(8-6-10)26(23,24)25;/h4-8H,1,9H2,2-3H3,(H,23,24,25);/q;+1/p-1
SMILES [Na+].CN1C(=O)N(CC=C)C(=O)c2c1nc(c3ccc(cc3)S(=O)(=O)[O-])n2C
IUPAC Name sodium;4-(3,7-dimethyl-2,6-dioxo-1-prop-2-enylpurin-8-yl)benzenesulfonate
Molecular Weight (g/mol) 398.37
Formula Weight 398.0661
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.