Learn More
1-(5-Methyl-2-nitrophenoxy)-2-(2-nitrophenoxy)ethane, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(5-Methyl-2-nitrophenoxy)-2-(2-nitrophenoxy)ethane |
| CAS | 96315-08-1 |
| Molecular Formula | C15H14N2O6 |
| Quantity | 25 mg |
| Synonym | 4-Methyl-1-nitro-2-[2-(2-nitrophenoxy)ethoxy]benzene |
| InChI Formula | InChI=1S/C15H14N2O6/c1-11-6-7-13(17(20)21)15(10-11)23-9-8-22-14-5-3-2-4-12(14)16(18)19/h2-7,10H,8-9H2,1H3 |
| SMILES | CC1=CC(OCCOC2=C([N+]([O-])=O)C=CC=C2)=C([N+]([O-])=O)C=C1 |
| IUPAC Name | 4-methyl-1-nitro-2-(2-(2-nitrophenoxy)ethoxy)benzene |
| Molecular Weight (g/mol) | 318.28 |
| Formula Weight | 318.09 |