missing translation for 'onlineSavingsMsg'
Learn More

1,5-Hexadiyne, 50% in pentane

Product Code. 11303477
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11303477

Brand: Thermo Scientific Alfa Aesar 018720.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-16-0 | C6H6 | 78.114 g/mol

1,5-Hexadiyne is used in the preparation of bisdehydro[12]annulene and biphenylene. It is also used as an important starting material in the preparation of (5Z,9Z)-5,9-hexadecadienoic acid. Further, it is involved in the preparation of linear organotin polymers by reaction with organotin dihydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-16-0
Molecular Formula C6H6
Molecular Weight (g/mol) 78.114
MDL Number MFCD00014924
InChI Key YFIBSNDOVCWPBL-UHFFFAOYSA-N
Synonym 1,5-hexadiyne, dipropargyl, bipropargyl, hc#cch2ch2c#ch, 1,5-hexadiyne in pentane, hccch2ch2cch, acmc-1b142, 1,5-hexadiyne 6ci, 8ci, 9ci, 1,5-hexadiyne solution, 50 wt. % in pentane 1g
PubChem CID 69402
ChEBI CHEBI:37821
IUPAC Name hexa-1,5-diyne
SMILES C#CCCC#C

Specifications

Density 0.805 g/mL
Flash Point -2°C (29°F)
Quantity 10 g
UN Number UN3295
Solubility Information Immiscible with water.
Formula Weight 78.11
Concentration or Composition (by Analyte or Components) 50% in pentane
Physical Form Liquid
Chemical Name or Material 1,5-Hexadiyne

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.