missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine, TRC

Product Code. 30403557
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403557 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403557 Supplier Toronto Research Chemicals Supplier No. TRCF59150810MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine
CAS 928830-73-3
Molecular Formula C11 H13 F N4 O4
Quantity 10 mg
InChI Formula InChI=1S/C11H13FN4O4/c1-13-2-4-14(5-3-13)10-6-8(12)9(15(17)18)7-11(10)16(19)20/h6-7H,2-5H2,1H3
SMILES CN1CCN(CC1)c2cc(F)c(cc2[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name 1-(5-fluoro-2,4-dinitrophenyl)-4-methylpiperazine
Molecular Weight (g/mol) 284.2437
Formula Weight 284.0921
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.