missing translation for 'onlineSavingsMsg'
Learn More

1,5,9-Decatriene, cis + trans, 97%, Thermo Scientific™

Product Code. 11325289
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325289 25 mL 25mL
11315289 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325289 Supplier Thermo Scientific Alfa Aesar Supplier No. A19822.AC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13393-64-1
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00055625
InChI Key GMGYODAMPOELNZ-MDZDMXLPSA-N
Synonym 1,5,9-decatriene, 5e-deca-1,5,9-triene, 1,5,9-decatriene, mixture of cis and trans, e-1,5,9-decatriene, 5e-1,5,9-decatriene, 5e-1,5,9-decatriene #, 1,5,9-decatriene, cis + trans
PubChem CID 5362636
IUPAC Name (5E)-deca-1,5,9-triene
SMILES C=CCC\C=C\CCC=C

Specifications

Density 0.765
Boiling Point 54°C to 55°C (14 mmHg)
Flash Point 46°C (115°F)
Refractive Index 1.448
Quantity 25 mL
UN Number UN3295
Formula Weight 136.24
Percent Purity 97%
Chemical Name or Material 1,5,9-Decatriene, cis + trans
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.