missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Pregnadien-11beta,17alpha,21-triol-3,20-dione-2,4,6,6,21,21-d6, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Prednisolone-2,4,6,6,21,21-D6 |
| Molecular Formula | C21 D6 H22 O5 |
| Quantity | 0.005 g |
| Synonym | 1,4-Pregnadien-11 beta,17 alpha,21-triol-3,20-dione-2,4,6,6,21,21-d6, (11β)-11,17,21-Trihydroxypregna-1,4-diene-3,20-dione-2,4,6,6,21,21-d6, [2,4,6,6,21,21-2H6]Prednisolone, Prednisolone-2,4,6,6,21,21-D6, Prednisolone-2,4,6,6,21,21-d6, Prednisolone-d6 |
| InChI Formula | InChI=1 S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26 H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1/i3D2,5 D,9 D,11D2 |
| SMILES | [2 H]C1=C[C@]2(C)[C@H]3 [C@@H](O)C[C@@]4(C)[C@@H](CC[C@]4(O)C(=O)C([2 H])([2 H])O)[C@@H]3 CC([2 H])([2 H])C2=C([2 H])C1=O |
| IUPAC Name | (8 S,9 S,10 R,11 S,13 S,14 S,17 R)-2,4,6,6-tetradeuterio-17-(2,2-dideuterio-2-hydroxyacetyl)-11,17-dihydroxy-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 366.481 |
| Formula Weight | 366.2313 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction