Learn More
1,4-Phenylenediamine, TRC
Product Code. 30011782
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011782 5 g 5g
30040800 1 g 1g
30009338 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011782 Supplier Toronto Research Chemicals Supplier No. TRCP3198455G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106-50-3
Molecular Formula C6 H8 N2
Molecular Weight (g/mol) 108.14
Synonym 1,4-Benzenediamine, p-Phenylenediamine (8CI), (4-Aminophenyl)amine, 1,4-Diaminobenzene, 1,4-Diaminobenzol, 1,4-Phenylenediamine, 4-Aminoaniline, 4-Phenylenediamine, BASF Ursol D, Benzofur D, Black for Fur D, C.I. 76060, C.I. Developer 13, C.I. Oxidation Base 10, Developer PF, Durafur Black R, Fouramine D, Fourrine 1, Fourrine D, Fur Black 41867, Fur Brown 41866, Fur Yellow, Furro D, Futramine D, NSC 4777, Nako H, Orsin, PDA, PPD, Paramine, Pelagol D, Pelagol DR, Pelagol Grey D, Peltol D, Renal PF, Rodol D, Rodol DJ, Tertral D, Ursol D, Zoba Black D, p-Aminoaniline, p-Benzenediamine, p-Diaminobenzene, pPDA
IUPAC Name benzene-1,4-diamine
SMILES Nc1ccc(N)cc1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C6H8N2/c7-5-1-2-6(8)4-3-5/h1-4H,7-8H2
Formula Weight 108.0687
Percent Purity >95%
Chemical Name or Material 1,4-Phenylenediamine (1,4-Diaminobenzene)
Grade HPLC