Learn More
1-(4-Methylphenyl)-1-cyclopropanecarboxylic Acid, TRC
Product Code. 30475187
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475187 50 mg 50mg
30465460 100 mg 100mg
30480162 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30475187 Supplier Toronto Research Chemicals Supplier No. TRCM21943550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83846-66-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.21
Synonym 1-(4-Methylphenyl)cyclopropanecarboxylic Acid, 1-(4-Methylphenyl)cyclopropane-1-carboxylic Acid, 1-p-Tolylcyclopropanecarboxylic Acid, NSC 156080
IUPAC Name 1-(4-methylphenyl)cyclopropane-1-carboxylic acid
SMILES Cc1ccc(cc1)C2(CC2)C(=O)O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C11H12O2/c1-8-2-4-9(5-3-8)11(6-7-11)10(12)13/h2-5H,6-7H2,1H3,(H,12,13)
Formula Weight 176.0837
Chemical Name or Material 1-(4-Methylphenyl)-1-cyclopropanecarboxylic Acid