Learn More
1-(4-Methoxyphenyl)-2-pyrrolidinone, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(4-Methoxyphenyl)-2-pyrrolidinone |
| CAS | 30425-47-9 |
| Molecular Formula | C11H13NO2 |
| Quantity | 50 mg |
| Synonym | 1-(p-Methoxyphenyl)-2-pyrrolidinone, N-(4-(Methoxyphenyl)pyrrolidin-2-one |
| InChI Formula | InChI=1S/C11H13NO2/c1-14-10-6-4-9(5-7-10)12-8-2-3-11(12)13/h4-7H,2-3,8H2,1H3 |
| SMILES | COc1ccc(cc1)N2CCCC2=O |
| IUPAC Name | 1-(4-methoxyphenyl)pyrrolidin-2-one |
| Molecular Weight (g/mol) | 191.23 |
| Formula Weight | 191.0946 |