Learn More
1,4-Dipiperazino-2,3,5,6-tetrafluorobenzene, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-Dipiperazino-2,3,5,6-tetrafluorobenzene |
| CAS | 502616-02-6 |
| Molecular Formula | C14H18F4N4 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C14H18F4N4/c15-9-11(17)14(22-7-3-20-4-8-22)12(18)10(16)13(9)21-5-1-19-2-6-21/h19-20H,1-8H2 |
| SMILES | OC(=O)COc1c(Cl)cc(Cl)c(Cl)n1 |
| Molecular Weight (g/mol) | 318.31 |