missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxynaphthalene, 96%

Product Code. 11418046
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11418046

Brand: Thermo Scientific Alfa Aesar A14095.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10075-62-4 | C12H12O2 | 188.226 g/mol

1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10075-62-4
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD00052378
InChI Key FWWRTYBQQDXLDD-UHFFFAOYSA-N
PubChem CID 82337
IUPAC Name 1,4-dimethoxynaphthalene
SMILES COC1=CC=C(C2=CC=CC=C21)OC

Specifications

Melting Point 84°C to 85°C
Quantity 5 g
Beilstein 2048082
Synonym naphthalene, 1,4-dimethoxy, 1,4-dimethoxy-naphthalene, maybridge1_003006, naphthalene,4-dimethoxy, acmc-1c6fs, 1,4-dimethoxy naphthalene, naphthalene,1,4-dimethoxy, 1,4-dimethoxynaphthalene, 1,4-dimethoxynaphthalene, for fluorescence, fluorescent marker hplc
Solubility Information Soluble in water.
Formula Weight 188.23
Percent Purity 96%
Chemical Name or Material 1,4-Dimethoxynaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.