Learn More
1,4-Dihydro-3-nitro-4-oxo-imidazo[5,1-c][1,2,4]triazine-8-carboxamide, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-Dihydro-3-nitro-4-oxo-imidazo[5,1-c][1,2,4]triazine-8-carboxamide |
| CAS | 287964-59-4 |
| Molecular Formula | C6 H4 N6 O4 |
| Quantity | 10 mg |
| Synonym | 1,4-Dihydro-3-nitro-4-oxo-imidazo[5,1-c][1,2,4]triazine-8-carboxamide, Imidazo[5,1-c][1,2,4]triazine-8-carboxamide, 1,4-dihydro-3-nitro-4-oxo- |
| InChI Formula | InChI=1S/C6H4N6O4/c7-3(13)2-4-9-10-5(12(15)16)6(14)11(4)1-8-2/h1,9H,(H2,7,13) |
| SMILES | NC(=O)c1ncn2C(=O)C(=NNc12)[N+](=O)[O-] |
| IUPAC Name | 3-nitro-4-oxo-1H-imidazo[5,1-c][1,2,4]triazine-8-carboxamide |
| Molecular Weight (g/mol) | 224.134 |
| Formula Weight | 224.029 |